Learn More
Fisherbrand™ Droplet Sample Storage Tray
Catalog No. 14127645 Shop All Fisherbrand Products
Change view
Click to view available options
Color:
Blue/White
Catalog No. Color
14-127-645 Blue/White
1 options
Catalog No. 14-127-645
Add to Cart
Add to Cart
Sample storage tray built for busy labs and clinical spaces.
  • Bin and tray are made from polystyrene
  • Lightweight and sturdy to hold up to 24 full specimen containers (Specimen containers not included)
  • Stackable with other trays to support as many as four high
  • Can be easily cleaned with common disinfectants and germicides
  • Removable Insert allows outer shell to be used for storage or transportation of laboratory products
  • Integrated label holder
Specifications
Autoclavable Not Autoclavable
Color Blue/White
Holds Specimen Containers
Material Polystyrene
Positions 24
Shape Rectangular
Stackable Yes
Sterility Non-sterile
Dimensions (L x W x H) 18.9 x 13 x 3.3 in.
Product Type BioTransport Carrier