Learn More
Biotium Drop-n-Stain EverBrite™ Mounting Medium with DAPI
Catalog No. 501964405
Encompass_Preferred
Drop-n-Stain EverBrite™ Mounting Medium with DAPI is wet-set antifade in a convenient dropper bottle for one step mounting and counterstaining. EverBrite™ protects fluorescent dyes from photobleaching, and is compatible with cyanine-based dyes like Cy dyes and Alexa Fluor 647. Unit size: 10 mL.
Catalog No. 50-196-4405 Supplier Biotium Supplier No. 23009
Drop-n-Stain EverBrite™ Mounting Medium with DAPI is wet-set antifade in a convenient dropper bottle for one step mounting and counterstaining. EverBrite™ protects fluorescent dyes from photobleaching, and is compatible with cyanine-based dyes like Cy dyes and Alexa Fluor 647. Unit size: 10 mL.