Learn More
Research Products International Corp Dropper Bulb, Natural Latex Bulb, 2 ml, 72 per Case
SDP
Catalog No. 50999149
Encompass
Dropper bulbs made from natural latex rubber for use with all small pipettes and Pasteur pipettes. Attach easily to pipettes for controlled dropping of liquids. Available to fit 2 ml pipettes and packaged 72 droppers per case. Dropper bulbs can be used with Disposable Plastic Pasteur Pipets also available from RPI.
Catalog No. 50-999-149 Supplier Research Products International Corp Supplier No. 142822
Dropper bulbs made from natural latex rubber for use with all small pipettes and Pasteur pipettes. Attach easily to pipettes for controlled dropping of liquids. Available to fit 2 ml pipettes and packaged 72 droppers per case. Dropper bulbs can be used with Disposable Plastic Pasteur Pipets also available from RPI.