Learn More
Hach Company Dropper, polyethylene, 1.0 mL graduated volume, 3.0 mL bulb draw capacity, 20/pk
Catalog No. 501996617 Shop All Hach Company Products
Encompass
Disposable, see-through transfer pipets. Made of unbreakable LPDE. Inert to biological fluids and most acids. Can be sealed, refrigerated, or gas sterilized.
Catalog No. 50-199-6617 Supplier Hach Company Supplier No. 2749220
Disposable, see-through transfer pipets. Made of unbreakable LPDE. Inert to biological fluids and most acids. Can be sealed, refrigerated, or gas sterilized.