Learn More
Unchained Labs High Lunatic Chips, OD 200. (25/box)
SDP
Catalog No. NC1205613
Encompass
12 boxes of 25 High Lunatic Chips, 0.03-200 OD, 16 samples per chip. For use with Little Lunatic or DropSense16. (Previously product code 1-230300-200)
Catalog No. NC1205613 Supplier Unchained Labs Supplier No. 7012008
12 boxes of 25 High Lunatic Chips, 0.03-200 OD, 16 samples per chip. For use with Little Lunatic or DropSense16. (Previously product code 1-230300-200)