Learn More
Unchained Labs Lunatic Verification Standard
SDP
Catalog No. 501125943
Encompass
Potassium Dichromate Kit (1ml verification standard, 3 glass vials). Used as verification standard for all Lunatic and DropSense platforms. (Previously product code 1-301010)
Catalog No. 50-112-5943 Supplier Unchained Labs Supplier No. 7012018
Potassium Dichromate Kit (1ml verification standard, 3 glass vials). Used as verification standard for all Lunatic and DropSense platforms. (Previously product code 1-301010)