Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HTL-Strefa DropSafe™ Safety Pen Needle
SDP

Catalog No. 22192017
Change view
Click to view available options
Needle Gauge:
31 G
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Needle Gauge
22-192-017 31 G
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 22-192-017 Supplier HTL-Strefa Supplier No. 8162
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

DropSafe Safety Pen Needles are sterile, single-use safety needles intended for use with pen injector devices for the injection of drugs. Passive safety technology is designed to improve safety for clinicians and patients.

  • Safer Drug Delivery for Clinicians & Patients
    • Viewing window
    • Red lock-out indicator
    • Primary cap used for backend protection
    • Ready for disposal to reduce a needle stick injury
  • Safety Pen Needles are recommended by Clinicians and Patients
    • When considering dose preparation and administration, 77% of nurses were confident in dose accuracy with a safety pen needle & insulin pen over an insulin syringe.
    • With an unobstructed needle viewing window, priming confirmation and injection site administration are clearly visible with DropSafe safety pen needle.
    • When a safety pen needle was used in the hospital and patients continued to use insulin pens with a pen needle at home, they will likely continue to be adherent, leading to a reduction of in-hospital costs. Patient satisfaction improved by 498% when able to continue taking insulin at home with the use of a pen needle versus a syringe.

Specifications

Color Purple
Needle Gauge 31 G
Product Type Safety Pen Needle
Quantity 100/Box
Length (Metric) Needle 6 mm
For Use With (Equipment) Insulin Injector Pen
Sterility Sterile
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.