Learn More
LabScientific Drosophila agar 80-100 mesh, 10 kg, powder, low gelling temp & high gel strength.
Catalog No. NC9349177
Encompass
Drosophila agar 80-100 mesh, 10 kg. Powder. A High Quality Microbiological Agar ~ primarily used in labs as a culture medium in Petri dishes. Gelidium agar has a low gelling temperature (34 to 36 degrees Celsius). It doesn't contain trace metals, soluble carbohydrates, carbohydrates or any spores that would inhibit bacterial growth, making it an exceptional medium for research projects. Gelidium is extracted from the species of red algae Rhodophyceae type form Gelidium Genera. Gelidium is soluble in boiling water, Kosher Certified, and classified as GRAS by the FDA. Melting point 88 C.
Catalog No. NC9349177 Supplier LabScientific Supplier No. FLY802010
Drosophila agar 80-100 mesh, 10 kg. Powder. A High Quality Microbiological Agar ~ primarily used in labs as a culture medium in Petri dishes. Gelidium agar has a low gelling temperature (34 to 36 degrees Celsius). It doesn't contain trace metals, soluble carbohydrates, carbohydrates or any spores that would inhibit bacterial growth, making it an exceptional medium for research projects. Gelidium is extracted from the species of red algae Rhodophyceae type form Gelidium Genera. Gelidium is soluble in boiling water, Kosher Certified, and classified as GRAS by the FDA. Melting point 88 C.