Learn More
Alkali Scientific ASI™ Drosophila Bottle Plugs, Recycled Polyester, 38.10mm x 31mm, 1000/Case
SDP
Catalog No. 503027324
Encompass_Preferred
These Drosophila plugs, made from recycled polyester with a bonded dense weave, are specifically designed for use with narrow bottles. The material provides an excellent barrier against mite infestation while offering superior breathability to maintain an optimal environment for your specimens. Reduced evaporation keeps food fresh longer, supporting healthier cultures. Unlike cellulose acetate, these plugs do not retain moisture in humid conditions, ensuring consistent performance in all environments. Precision-engineered using proprietary dies, these plugs ensure a consistent fit with minimal waste. Their strong, durable construction eliminates clinging fibers and prevents breakage, making them a superior alternative to traditional options. Proudly made in the USA, these plugs are an excellent choice for laboratories seeking high-quality closures for narrow bottles. Packaged in a case of 1000, they are ideal for high-volume use.
Catalog No. 50-302-7324 Supplier Alkali Scientific Supplier No. DF30PB
These Drosophila plugs, made from recycled polyester with a bonded dense weave, are specifically designed for use with narrow bottles. The material provides an excellent barrier against mite infestation while offering superior breathability to maintain an optimal environment for your specimens. Reduced evaporation keeps food fresh longer, supporting healthier cultures. Unlike cellulose acetate, these plugs do not retain moisture in humid conditions, ensuring consistent performance in all environments. Precision-engineered using proprietary dies, these plugs ensure a consistent fit with minimal waste. Their strong, durable construction eliminates clinging fibers and prevents breakage, making them a superior alternative to traditional options. Proudly made in the USA, these plugs are an excellent choice for laboratories seeking high-quality closures for narrow bottles. Packaged in a case of 1000, they are ideal for high-volume use.