Learn More
Percival Scientific Drosophila Incubator, DR36NL, 29.7 cu. ft., non-lighted, 6 tiers, additive pan-type humidifier, coated coils, 2-44° C ±0.5°, IntellusUltra controller
SDP
Catalog No. NC3793903
Encompass_Preferred
The DR36NL incubator is designed with features for optimal rearing of Drosophila melanogaster. Interior / exterior of smooth white galvanized steel with foamed-in-place non-CFC insulation and a stainless-steel floor for corrosion resistance. A phenolic coated coil prevents waste damage. Additive pan-type humidifier keeps media from drying out and maintains optimal humidity. Six white epoxy coated steel wire shelves adjustable in ½” increments. Self-contained air-cooled condensing unit with hot gas bypass system for continuous compressor operation extends life, ensuring precise temperature control by alternately cycling refrigerant and hot gas to coil; this also prolongs life of compressor, and eliminates risk of ice build up in coil. Top mounted refrigeration system allows easy access for maintenance and cleaning. Contact sales@percival-scientific.com for more options.
Catalog No. NC3793903 Supplier Percival Scientific Supplier No. DR36NL
Add to Cart
Add to Cart
Request a Quote
The DR36NL incubator is designed with features for optimal rearing of Drosophila melanogaster. Interior / exterior of smooth white galvanized steel with foamed-in-place non-CFC insulation and a stainless-steel floor for corrosion resistance. A phenolic coated coil prevents waste damage. Additive pan-type humidifier keeps media from drying out and maintains optimal humidity. Six white epoxy coated steel wire shelves adjustable in ½” increments. Self-contained air-cooled condensing unit with hot gas bypass system for continuous compressor operation extends life, ensuring precise temperature control by alternately cycling refrigerant and hot gas to coil; this also prolongs life of compressor, and eliminates risk of ice build up in coil. Top mounted refrigeration system allows easy access for maintenance and cleaning. Contact sales@percival-scientific.com for more options.