Learn More
Percival Scientific Drosophila Incubator, DR41VLH3, 37.2 cu. ft., vertical IncuWhite LEDs, 5 tiers, pan-type hum., dehum. and standard sensor, coated coils, 5-44° C ±0.5°, IntellusUltra controller
SDP
Catalog No. NC3788670
Encompass_Preferred
The DR41VLH3 incubator is designed with features for optimal rearing of Drosophila melanogaster. IncuWhite LEDs are dimmable between 5-100% output with up to 120 µmoles/m2/s irradiance @ 6” from the LEDs. Interior / exterior of smooth white galvanized steel with foamed-in-place non-CFC insulation and a stainless-steel floor for corrosion resistance. A phenolic coated coil prevents waste damage. Pan-type humidifier and dehumidifier keep media from drying out and maintains optimal humidity. Five white epoxy coated steel wire shelves adjustable in ½” increments. Self-contained air-cooled condensing unit with hot gas bypass system for continuous compressor operation extends life, ensuring precise temperature control by alternately cycling refrigerant and hot gas to coil; this also prolongs life of compressor, and eliminates risk of ice build up in coil. Top mounted refrigeration system allows easy access for maintenance and cleaning. Contact sales@percival-scientific.com for more options.
Catalog No. NC3788670 Supplier Percival Scientific Supplier No. DR41VLH3
Add to Cart
Add to Cart
Request a Quote
The DR41VLH3 incubator is designed with features for optimal rearing of Drosophila melanogaster. IncuWhite LEDs are dimmable between 5-100% output with up to 120 µmoles/m2/s irradiance @ 6” from the LEDs. Interior / exterior of smooth white galvanized steel with foamed-in-place non-CFC insulation and a stainless-steel floor for corrosion resistance. A phenolic coated coil prevents waste damage. Pan-type humidifier and dehumidifier keep media from drying out and maintains optimal humidity. Five white epoxy coated steel wire shelves adjustable in ½” increments. Self-contained air-cooled condensing unit with hot gas bypass system for continuous compressor operation extends life, ensuring precise temperature control by alternately cycling refrigerant and hot gas to coil; this also prolongs life of compressor, and eliminates risk of ice build up in coil. Top mounted refrigeration system allows easy access for maintenance and cleaning. Contact sales@percival-scientific.com for more options.