Learn More
Active Motif Drosophila Negative Control Primer Set 1
SDP
Catalog No. NC1353113
Encompass
Model Organism Control qPCR Primer Sets were designed to serve as positive or negative ChIP controls when performing chromatin immunoprecipitation with samples from rat, zerbrafish, Drosophila or budding yeast. Each primer set is a mixture of forward and reverse primers that have been validated for qPCR and endpoint PCR of ChIP DNA from multiple samples.
Catalog No. NC1353113 Supplier Active Motif Supplier No. 71028
Model Organism Control qPCR Primer Sets were designed to serve as positive or negative ChIP controls when performing chromatin immunoprecipitation with samples from rat, zerbrafish, Drosophila or budding yeast. Each primer set is a mixture of forward and reverse primers that have been validated for qPCR and endpoint PCR of ChIP DNA from multiple samples.