Learn More
Active Motif Drosophila Positive Control Primer Set Pbgs
SDP
Catalog No. NC1353110
Encompass
Model Organism Control qPCR Primer Sets were designed to serve as positive or negative ChIP controls when performing chromatin immunoprecipitation with samples from rat, zerbrafish, Drosophila or budding yeast. Each primer set is a mixture of forward and reverse primers that have been validated for qPCR and endpoint PCR of ChIP DNA from multiple samples.
Catalog No. NC1353110 Supplier Active Motif Supplier No. 71037
Model Organism Control qPCR Primer Sets were designed to serve as positive or negative ChIP controls when performing chromatin immunoprecipitation with samples from rat, zerbrafish, Drosophila or budding yeast. Each primer set is a mixture of forward and reverse primers that have been validated for qPCR and endpoint PCR of ChIP DNA from multiple samples.