Learn More
Thomas Scientific Drosophila Vial, Narrow, Polystyrene, 25 x 95mm, Tray
SDP
Catalog No. 501459288
Encompass
Drosophila Vials Made of premium grade polystyrene Glass-like clarity for sample observation Disposable vials are a safe and cost-saving alternative to glass vials Narrow or wide vials available Both narrow and wide vials are available in bulk or tray packaging Tray packs are great for quick, effortless filling of vials Drosophila Plugs Made of high density foam Offer minimal shedding and reduces evaporation Maintains tight fit after repeated removal and plugging
Catalog No. 50-145-9288 Supplier Thomas Scientific Supplier No. 1159Q82
Drosophila Vials Made of premium grade polystyrene Glass-like clarity for sample observation Disposable vials are a safe and cost-saving alternative to glass vials Narrow or wide vials available Both narrow and wide vials are available in bulk or tray packaging Tray packs are great for quick, effortless filling of vials Drosophila Plugs Made of high density foam Offer minimal shedding and reduces evaporation Maintains tight fit after repeated removal and plugging