Learn More
CELLTREAT Scientific Products Drosophila Vial, Wide Opening, Polystyrene, Tray, Non-sterile
SDP
Catalog No. 5000144318
Encompass_Preferred
Suitable for drosophila-based applications, including studies of genetics, developmental biology, pharmacology, disease modeling, behavior, and lifespanCrystal-clear polystyrene vials provide a controlled environment for fruit flies to feed, mate, and lay eggsDesigned with a straight wall and flat bottom for easy filling, plugging, and racking in traysSafe, lighter, and more economical alternative to traditional glass vialsAvailable in bulk packs and convenient tray packs
Catalog No. 50-001-44318 Supplier CELLTREAT Scientific Products Supplier No. 229901
Suitable for drosophila-based applications, including studies of genetics, developmental biology, pharmacology, disease modeling, behavior, and lifespanCrystal-clear polystyrene vials provide a controlled environment for fruit flies to feed, mate, and lay eggsDesigned with a straight wall and flat bottom for easy filling, plugging, and racking in traysSafe, lighter, and more economical alternative to traditional glass vialsAvailable in bulk packs and convenient tray packs