Learn More
Cayman Chemical DrospIrenon 10mg
SDP
Catalog No. 502993873
Encompass_Preferred
A synthetic progestogen; binds to the progesterone, mineralocorticoid, and androgen receptors with binding affinities of 20, 230, and 65% relative to R5020, aldosterone, and R1881, respectively; inhibits spontaneous ovulation in rats (ID50s = 0.3-1.0 mg/day); maintains pregnancy in ovariectomized pregnant rats; reduces serum testosterone and luteinizing hormone in cynomolgus monkeys; inhibits testosterone-induced growth of the seminal vesicles and prostate in castrated rats
Catalog No. 50-299-3873 Supplier Cayman Chemical Supplier No. 2334710 MG
A synthetic progestogen; binds to the progesterone, mineralocorticoid, and androgen receptors with binding affinities of 20, 230, and 65% relative to R5020, aldosterone, and R1881, respectively; inhibits spontaneous ovulation in rats (ID50s = 0.3-1.0 mg/day); maintains pregnancy in ovariectomized pregnant rats; reduces serum testosterone and luteinizing hormone in cynomolgus monkeys; inhibits testosterone-induced growth of the seminal vesicles and prostate in castrated rats