Learn More
Medchemexpress LLC Drospirenone-d4 | 2376035-94-6 | 98.5% | C24H26D4O3 | 2.5 MG
SDP
Catalog No. 5000432289
Encompass_Preferred
Drospirenone-d4 is the deuterium labeled Drospirenone, a synthetic progestin that is an analog to spironolactone. Stable heavy isotopes are incorporated into drug molecules, primarily as tracers for quantitation during drug development. Deuteration has the potential to influence the pharmacokinetic and metabolic profiles of drugs. This compound is for research use only.
  • Used as a tracer
  • Used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
Catalog No. 50-004-32289 Supplier Medchemexpress LLC Supplier No. HYB0111S2.5MG
Drospirenone-d4 is the deuterium labeled Drospirenone, a synthetic progestin that is an analog to spironolactone. Stable heavy isotopes are incorporated into drug molecules, primarily as tracers for quantitation during drug development. Deuteration has the potential to influence the pharmacokinetic and metabolic profiles of drugs. This compound is for research use only.
  • Used as a tracer
  • Used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS