Learn More
Cayman Chemical Drotaverine (hydrochloride), Purity: ≥98%, CAS: 985-12-6, 10 mg
SDP
Catalog No. 502982746
Encompass_Preferred
An alkaloid that has been described as an inhibitor of phosphodiesterase 4 and negative allosteric modulator of L-type Ca2+ channels
Catalog No. 50-298-2746 Supplier Cayman Chemical Supplier No. 2094410 MG
An alkaloid that has been described as an inhibitor of phosphodiesterase 4 and negative allosteric modulator of L-type Ca2+ channels