Learn More
Cayman Chemical DrotaverInhydrochlorIde 50mg
SDP
Catalog No. 502981582
Encompass_Preferred
An alkaloid that has been described as an inhibitor of phosphodiesterase 4 and negative allosteric modulator of L-type Ca2+ channels
Catalog No. 50-298-1582 Supplier Cayman Chemical Supplier No. 2094450 MG
An alkaloid that has been described as an inhibitor of phosphodiesterase 4 and negative allosteric modulator of L-type Ca2+ channels