Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Droxidopa | 23651-95-8 | MFCD00799030 | 98.8% | C9H11NO5 | 10MG
SDP

Catalog No. 5000260050
Encompass_Preferred

Droxidopa (L-DOPS) is an orally active precursor of norepinephrine used in research to evaluate orthostatic hypotension, noradrenergic pathways, and related pharmacology. Supplied as an analytical-grade reagent for biochemical, pharmacological, and assay-development applications.

  • Orally active norepinephrine precursor for research use.
  • High purity suitable for analytical and pharmacological studies.
  • Chemical formula C9H11NO5; molecular weight 213.19 g·mol-1.
  • CAS 23651-95-8 for unambiguous identification.
  • Packaged in small sizes convenient for assay development and standards.

Catalog No. 50-002-60050 Supplier Medchemexpress LLC Supplier No. HY1345810MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Droxidopa (L-DOPS) is an orally active precursor of norepinephrine used in research to evaluate orthostatic hypotension, noradrenergic pathways, and related pharmacology. Supplied as an analytical-grade reagent for biochemical, pharmacological, and assay-development applications.

  • Orally active norepinephrine precursor for research use.
  • High purity suitable for analytical and pharmacological studies.
  • Chemical formula C9H11NO5; molecular weight 213.19 g·mol-1.
  • CAS 23651-95-8 for unambiguous identification.
  • Packaged in small sizes convenient for assay development and standards.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.