Learn More
Medchemexpress LLC HY-13458 5mg Medchemexpress, Droxidopa CAS:23651-95-8 Purity:>98%
SDP
Catalog No. 501969854
Encompass_Preferred
Medchemexpress, HY-13458 5mg Droxidopa CAS:23651-95-8 Droxidopa(L-DOPS), the mixture of Droxidopa (w/w80%) and Pharmaceutical starch (w/w20%), acts as a prodrug to the neurotransmitters norepinephrine (noradrenaline) and epinephrine (adrenaline); Droxidopa(L-DOPS) is capable of crossing the protective blood–brain barrier. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-9854 Supplier Medchemexpress LLC Supplier No. HY134585MG
Medchemexpress, HY-13458 5mg Droxidopa CAS:23651-95-8 Droxidopa(L-DOPS), the mixture of Droxidopa (w/w80%) and Pharmaceutical starch (w/w20%), acts as a prodrug to the neurotransmitters norepinephrine (noradrenaline) and epinephrine (adrenaline); Droxidopa(L-DOPS) is capable of crossing the protective blood–brain barrier. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.