Learn More
TARGETMOL CHEMICALS INC DROXINOSTAT 25MG
SDP
Catalog No. 502579353
Encompass_Preferred
Also available in 2 mg 5 mg 10 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Droxinostat (NS 41080) is a selective inhibitor of HDAC mostly for HDACs 6 and 8 with IC50 of 2.47 uM and 1.46 uM greater than 8-fold selective against HDAC3 and no inhibition to HDAC1 2 4 5 7 9 and 10. purity: 99%
Catalog No. 50-257-9353 Supplier TARGETMOL CHEMICALS INC Supplier No. T648125MG
Also available in 2 mg 5 mg 10 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Droxinostat (NS 41080) is a selective inhibitor of HDAC mostly for HDACs 6 and 8 with IC50 of 2.47 uM and 1.46 uM greater than 8-fold selective against HDAC3 and no inhibition to HDAC1 2 4 5 7 9 and 10. purity: 99%