Learn More
ABclonal Technology DRP1 Rabbit mAb
SDP
Catalog No. 5000005714
Encompass_Preferred
This gene encodes a member of the dynamin superfamily of GTPases The encoded protein mediates mitochondrial and peroxisomal division and is involved in developmentally regulated apoptosis and programmed necrosis Dysfunction of this gene is implicated in several neurological disorders including Alzheimer s disease Mutations in this gene are associated with the autosomal dominant disorder encephalopathy lethal due to defective mitochondrial and peroxisomal fission (EMPF) Alternative splicing results in multiple transcript variants encoding different isoforms
Catalog No. 50-000-05714 Supplier ABclonal Technology Supplier No. A2196820UL
This gene encodes a member of the dynamin superfamily of GTPases The encoded protein mediates mitochondrial and peroxisomal division and is involved in developmentally regulated apoptosis and programmed necrosis Dysfunction of this gene is implicated in several neurological disorders including Alzheimer s disease Mutations in this gene are associated with the autosomal dominant disorder encephalopathy lethal due to defective mitochondrial and peroxisomal fission (EMPF) Alternative splicing results in multiple transcript variants encoding different isoforms