Learn More
Medchemexpress LLC DRP1i27 dihydrochloride | 1453028-33-5 | C20H28Cl2N6O | 100 MG
SDP
Catalog No. 5000373678
Encompass_Preferred
DRP1i27 dihydrochloride is a potent inhibitor of human Drp1 (dynamin-related protein 1). It binds to the GTPase site of Drp1, forming hydrogen bonds with Gln34 and Asp218. This compound targets Drp1-mediated mitochondrial fission in cell line models and offers protection against simulated ischemia-reperfusion injury.
  • Directly binds to and inhibits the GTPase activity of human Drp1 (0-50 μM).
  • Increases cellular networks of mitochondria in human and mouse fibroblasts in a Drp1-dependent manner.
  • Exhibits a binding affinity of 286 μM in the SPR assay and a KD value of 190 μM via the MST assay.
Catalog No. 50-003-73678 Supplier Medchemexpress LLC Supplier No. HY152086A100MG
DRP1i27 dihydrochloride is a potent inhibitor of human Drp1 (dynamin-related protein 1). It binds to the GTPase site of Drp1, forming hydrogen bonds with Gln34 and Asp218. This compound targets Drp1-mediated mitochondrial fission in cell line models and offers protection against simulated ischemia-reperfusion injury.
  • Directly binds to and inhibits the GTPase activity of human Drp1 (0-50 μM).
  • Increases cellular networks of mitochondria in human and mouse fibroblasts in a Drp1-dependent manner.
  • Exhibits a binding affinity of 286 μM in the SPR assay and a KD value of 190 μM via the MST assay.