Learn More
Medchemexpress LLC DRP1i27 dihydrochloride | C20H28Cl2N6O | 1 ML
SDP
Catalog No. 5000373694
Encompass_Preferred
DRP1i27 dihydrochloride is a potent inhibitor of human Drp1 (dynamin-related protein 1). It binds to the GTPase site of Drp1, forming hydrogen bonds with Gln34 and Asp218. This compound targets Drp1-mediated mitochondrial fission in cell line models and offers protection against simulated ischemia-reperfusion injury.
  • Directly binds to and inhibits the GTPase activity of human Drp1.
  • Increases cellular networks of mitochondria in human and mouse fibroblasts in a Drp1-dependent manner.
  • Has a binding affinity of 286 μM in the SPR assay and a KD value of 190 μM via the MST assay.
Catalog No. 50-003-73694 Supplier Medchemexpress LLC Supplier No. HY152086A10MM/1ML
DRP1i27 dihydrochloride is a potent inhibitor of human Drp1 (dynamin-related protein 1). It binds to the GTPase site of Drp1, forming hydrogen bonds with Gln34 and Asp218. This compound targets Drp1-mediated mitochondrial fission in cell line models and offers protection against simulated ischemia-reperfusion injury.
  • Directly binds to and inhibits the GTPase activity of human Drp1.
  • Increases cellular networks of mitochondria in human and mouse fibroblasts in a Drp1-dependent manner.
  • Has a binding affinity of 286 μM in the SPR assay and a KD value of 190 μM via the MST assay.