Learn More
Medchemexpress LLC Drp1i27 (dihydrochloride) | C20H28Cl2N6O | 25 MG
SDP
Catalog No. 5000374463
Encompass_Preferred
DRP1i27 dihydrochloride is a potent inhibitor of human Drp1 (dynamin-related protein 1). It binds to the GTPase site of Drp1 through hydrogen bonds to Gln34 and Asp218. This product targets Drp1-mediated mitochondrial fission in cell line models and offers protection against simulated ischemia-reperfusion injury.
  • Potent inhibitor of human Drp1 (dynamin-related protein 1)
  • Binds to the GTPase site of Drp1
  • Targets Drp1-mediated mitochondrial fission in cell line models
  • Protects against simulated ischemia-reperfusion injury
  • Increases cellular networks of mitochondria in human and mouse fibroblasts in a Drp1-dependent manner
Catalog No. 50-003-74463 Supplier Medchemexpress LLC Supplier No. 25MGHY152086A
DRP1i27 dihydrochloride is a potent inhibitor of human Drp1 (dynamin-related protein 1). It binds to the GTPase site of Drp1 through hydrogen bonds to Gln34 and Asp218. This product targets Drp1-mediated mitochondrial fission in cell line models and offers protection against simulated ischemia-reperfusion injury.
  • Potent inhibitor of human Drp1 (dynamin-related protein 1)
  • Binds to the GTPase site of Drp1
  • Targets Drp1-mediated mitochondrial fission in cell line models
  • Protects against simulated ischemia-reperfusion injury
  • Increases cellular networks of mitochondria in human and mouse fibroblasts in a Drp1-dependent manner