Learn More
Medchemexpress LLC DRP1i27 dihydrochloride | C20H28Cl2N6O | 5 MG
SDP
Catalog No. 5000373598
Encompass_Preferred
DRP1i27 dihydrochloride is a potent inhibitor of human Drp1 (dynamin-related protein 1). It binds to the GTPase site of Drp1, forming hydrogen bonds with Gln34 and Asp218. This compound targets Drp1-mediated mitochondrial fission in cell line models and offers protection against simulated ischemia-reperfusion injury.
  • Directly binds to and inhibits the GTPase activity of human Drp1
  • Increases cellular networks of mitochondria in human and mouse fibroblasts
  • Protective effects against simulated ischemia-reperfusion injury
Catalog No. 50-003-73598 Supplier Medchemexpress LLC Supplier No. HY152086A5MG
DRP1i27 dihydrochloride is a potent inhibitor of human Drp1 (dynamin-related protein 1). It binds to the GTPase site of Drp1, forming hydrogen bonds with Gln34 and Asp218. This compound targets Drp1-mediated mitochondrial fission in cell line models and offers protection against simulated ischemia-reperfusion injury.
  • Directly binds to and inhibits the GTPase activity of human Drp1
  • Increases cellular networks of mitochondria in human and mouse fibroblasts
  • Protective effects against simulated ischemia-reperfusion injury