Learn More
Med Vet International Surgical Drape 3MIoban2 Antimicrobial Incise Drape 23 W X 23 L inch Sterile, 6648Ez, 10/Box
Catalog No. 502829026
Encompass_Preferred
3M surgical drapes help reduce infection at the site of the wound They contain antimicrobial in the adhesive for added safety Made from low-memory film, they're flexible and conform to the body A full-width handle enables a quick release of the liners 3M Ioban 2 antimicrobial incise drapes provide a sterile surface that encompasses the entire wound 23 x 23 inches
Catalog No. 50-282-9026 Supplier Med Vet International Supplier No. 405682BX
3M surgical drapes help reduce infection at the site of the wound They contain antimicrobial in the adhesive for added safety Made from low-memory film, they're flexible and conform to the body A full-width handle enables a quick release of the liners 3M Ioban 2 antimicrobial incise drapes provide a sterile surface that encompasses the entire wound 23 x 23 inches