Learn More
Med Vet International McKesson Absorbent Gelling Fiber Dressing, 2 x 2 Inch, 87200, 100/Case
Catalog No. 503044932
Encompass_Preferred
Absorbent Gelling Fiber Dressing McKesson Carboxymethyl Cellulose (CMC) 2 X 2 Inch Highly absorbent Excellent wet and dry strength Supports moist wound environment for faster healing Low risk of maceration Quilting pattern 2 x 2 inches
Catalog No. 50-304-4932 Supplier Med Vet International Supplier No. 1138625CS
Absorbent Gelling Fiber Dressing McKesson Carboxymethyl Cellulose (CMC) 2 X 2 Inch Highly absorbent Excellent wet and dry strength Supports moist wound environment for faster healing Low risk of maceration Quilting pattern 2 x 2 inches