Learn More
Med Vet International Transparent Film Dressing DermaView II™ 2-3/7 X 2-3/4 Inch Frame Style Delivery Rectangle Sterile, 00252E, 100/Box
Catalog No. 503044343
Encompass_Preferred
Moisture-vapor permeable transparent dressing aids in the prevention of bacterial contamination Transparent for evaluation of wound progress Conformable: Picture frame border allows for easy application to hard-to-conform-to areas Waterproof 6 x 7 centimeters
Catalog No. 50-304-4343 Supplier Med Vet International Supplier No. 714363BX
Moisture-vapor permeable transparent dressing aids in the prevention of bacterial contamination Transparent for evaluation of wound progress Conformable: Picture frame border allows for easy application to hard-to-conform-to areas Waterproof 6 x 7 centimeters