Learn More
Med Vet International ComfortFoam™ Border Silicone Adhesive with Border Silicone Foam Dressing, 6 x 6 Inch, Sterile, 00318E, Each
Catalog No. 503043460
Encompass_Preferred
ComfortFoam border dressing is self-adherent for easy applications soft silicone and foam materials make the dressing highly absorbent and flexible Moisture-proof outer film allows for water evaporation, so the dressing stays dry, even after showering Silicone adhesive and a low-profile border hold the dressing in place without sticking to the wound bed, minimizing wound trauma Dressing helps wounds stay moist and thermally insulated for quicker healing DermaRite's foam dressing is suitable for days-long use on various types of ulcers and traumatic wounds
Catalog No. 50-304-3460 Supplier Med Vet International Supplier No. 835616EA
ComfortFoam border dressing is self-adherent for easy applications soft silicone and foam materials make the dressing highly absorbent and flexible Moisture-proof outer film allows for water evaporation, so the dressing stays dry, even after showering Silicone adhesive and a low-profile border hold the dressing in place without sticking to the wound bed, minimizing wound trauma Dressing helps wounds stay moist and thermally insulated for quicker healing DermaRite's foam dressing is suitable for days-long use on various types of ulcers and traumatic wounds