Learn More
Med Vet International Allevyn Gentle Border Silicone Foam Dressing, 7 x 7 Inch, Sterile, 66800280, Each
Catalog No. 503044201
Encompass_Preferred
Allevyn advanced wound dressings are easy to remove and reapply as needed Three technologies, including a soft silicone adhesive gel reduce wound trauma during bandage changes Allevyn gentle dressings with borders maintain fluid balance to promote faster healing and reduce pain They're made for wounds that have a full and partial-thickness, are shallow, chronic and acute, malignant and/or infected Allevyn wound dressings are suitable for protecting 1st- and 2nd-degree burns 17.5 x 17.5 centimeters
Catalog No. 50-304-4201 Supplier Med Vet International Supplier No. 665774EA
Allevyn advanced wound dressings are easy to remove and reapply as needed Three technologies, including a soft silicone adhesive gel reduce wound trauma during bandage changes Allevyn gentle dressings with borders maintain fluid balance to promote faster healing and reduce pain They're made for wounds that have a full and partial-thickness, are shallow, chronic and acute, malignant and/or infected Allevyn wound dressings are suitable for protecting 1st- and 2nd-degree burns 17.5 x 17.5 centimeters