Learn More
Med Vet International Foam Dressing Allevyn Life 6-1/16 X 6-1/16 Inch With Border Film Backing Silicone Gel Adhesive Quadrilobe Sterile, 66801069, 10/Box
Catalog No. 503043897
Encompass_Preferred
ALLEVYN LIFE is a silicone-gel adhesive, composite hydrocellular foam dressing Redistributes pressure more evenly than traditional foam dressing Up to 7 day wear time (except on the sacrum, which is up to a 5 day wear time) Indicated for shallow, granulating wounds, chronic and acute exudative wounds, full- and partial-thickness wounds such as pressure ulcers, leg ulcers, diabetic foot ulcers, infected wounds, malignant wounds, surgical wounds, first- and second-degree burns, donor sites, skin tears and fungating ulcers Multi-layered design provides cushioning and helps to redistribute pressure Silicone wound contact layer minimizes trauma to the wound and pain to the patient during dressing changes8 while allowing the dressing to be repositoned
Catalog No. 50-304-3897 Supplier Med Vet International Supplier No. 834487BX
ALLEVYN LIFE is a silicone-gel adhesive, composite hydrocellular foam dressing Redistributes pressure more evenly than traditional foam dressing Up to 7 day wear time (except on the sacrum, which is up to a 5 day wear time) Indicated for shallow, granulating wounds, chronic and acute exudative wounds, full- and partial-thickness wounds such as pressure ulcers, leg ulcers, diabetic foot ulcers, infected wounds, malignant wounds, surgical wounds, first- and second-degree burns, donor sites, skin tears and fungating ulcers Multi-layered design provides cushioning and helps to redistribute pressure Silicone wound contact layer minimizes trauma to the wound and pain to the patient during dressing changes8 while allowing the dressing to be repositoned