Learn More
Med Vet International Allevyn Gentle Border Lite Silicone Foam Dressing, Sterile, 66800834, 10/Box
Catalog No. 503044957
Encompass_Preferred
An Allevyn Gentle Border Lite dressing is safe for use on fragile and sensitive skin It's advanced technology sustains a moist environment to help wounds heal faster The foam core makes the dressing flexible and able to fit curved areas A showerproof silicone adhesive stays in place, even when wet and is gentle on skin during removal You can reposition the Allevyn wound dressing without losing adhesion It creates a bacterial barrier that lowers the chance of infection Each dressing lasts up to 1 week for fewer changes 7.5 x 7.5 centimeters
Catalog No. 50-304-4957 Supplier Med Vet International Supplier No. 767990BX
An Allevyn Gentle Border Lite dressing is safe for use on fragile and sensitive skin It's advanced technology sustains a moist environment to help wounds heal faster The foam core makes the dressing flexible and able to fit curved areas A showerproof silicone adhesive stays in place, even when wet and is gentle on skin during removal You can reposition the Allevyn wound dressing without losing adhesion It creates a bacterial barrier that lowers the chance of infection Each dressing lasts up to 1 week for fewer changes 7.5 x 7.5 centimeters