Learn More
Med Vet International Allevyn™ Gentle Border Lite Silicone Gel Adhesive with Border Thin Silicone Foam Dressing, 4 x 4 Inch, Sterile, 66800835, 10/Box
Catalog No. 503044975
Encompass_Preferred
The Allevyn Gentle Border Lite dressing is safe for use on fragile and sensitive skin Its advanced technology sustains a moist environment to help wounds heal faster The foam core makes the dressing flexible and able to fit curved areas A showerproof silicone adhesive stays in place, even when wet and is gentle on the skin during removal You can reposition the Allevyn wound dressing without losing adhesion It creates a bacterial barrier that lowers the chance of infection Each dressing lasts up to 1 week for fewer changes 10 x 10 centimeters
Catalog No. 50-304-4975 Supplier Med Vet International Supplier No. 767992BX
The Allevyn Gentle Border Lite dressing is safe for use on fragile and sensitive skin Its advanced technology sustains a moist environment to help wounds heal faster The foam core makes the dressing flexible and able to fit curved areas A showerproof silicone adhesive stays in place, even when wet and is gentle on the skin during removal You can reposition the Allevyn wound dressing without losing adhesion It creates a bacterial barrier that lowers the chance of infection Each dressing lasts up to 1 week for fewer changes 10 x 10 centimeters