Learn More
Med Vet International Foam Dressing Mepilex Border Flex 3 X 3 Inch With Border Film Backing Silicone Adhesive Square Sterile, 595200, 5/Box
Catalog No. 503044454
Encompass_Preferred
Proprietary Flex technology enables 360° stretch, thanks to patent-pending Y-cuts in the spreading and retention layers Mepilex Border Flex manages both normal and viscous exudate
Catalog No. 50-304-4454 Supplier Med Vet International Supplier No. 1114379BX
Proprietary Flex technology enables 360° stretch, thanks to patent-pending Y-cuts in the spreading and retention layers Mepilex Border Flex manages both normal and viscous exudate