Learn More
Med Vet International Mepilex Border Flex Lite Silicone Foam Dressing, 6 x 6in, 5/Box, 581500, 10 Boxes
Catalog No. 503044514
Encompass_Preferred
Thin Foam Dressing Mepilex Border Flex Lite 6 X 6 Inch With Border Film Backing Silicone Adhesive Square Sterile Designed to stay on and conform Four-layer dressing absorbs and channels exudate away from the wound bed and allows you to track exudate progress without disturbing the wound 6 x 6 inches
Catalog No. 50-304-4514 Supplier Med Vet International Supplier No. 1195998CS
Thin Foam Dressing Mepilex Border Flex Lite 6 X 6 Inch With Border Film Backing Silicone Adhesive Square Sterile Designed to stay on and conform Four-layer dressing absorbs and channels exudate away from the wound bed and allows you to track exudate progress without disturbing the wound 6 x 6 inches