Learn More
Med Vet International Foam Dressing Mepilex Border Post-Op 4 X 10 Inch With Border Film Backing Silicone Adhesive Rectangle Sterile, 496455, Each
Catalog No. 503044739
Encompass_Preferred
Design gives very high flexibility and makes it an excellent choice to be used over joints such as hips and knees Wound contact layer is perforated silicone and Safetac coated polyurethane film Transparent border allows inspection of peri-wound skin without removal Spreading layer made from nonwoven viscose and polyester
Catalog No. 50-304-4739 Supplier Med Vet International Supplier No. 1058163EA
Design gives very high flexibility and makes it an excellent choice to be used over joints such as hips and knees Wound contact layer is perforated silicone and Safetac coated polyurethane film Transparent border allows inspection of peri-wound skin without removal Spreading layer made from nonwoven viscose and polyester