Learn More
Med Vet International Thin Hydrocolloid Dressing DermaFilm 4 X 4 Inch Square With Border, 00259E, 10/Box
Catalog No. 503043766
Encompass_Preferred
Mild adhesive surface area adheres to peri-wound skin, but will not stick to wound bed Supports moist wound healing and autolytic debridement Provides thermal insulation and protection Comfortable and conformable Can be safely worn for several days, depending on wound condition and exudate levels 10.2 x 10.2 centimeters
Catalog No. 50-304-3766 Supplier Med Vet International Supplier No. 583419BX
Mild adhesive surface area adheres to peri-wound skin, but will not stick to wound bed Supports moist wound healing and autolytic debridement Provides thermal insulation and protection Comfortable and conformable Can be safely worn for several days, depending on wound condition and exudate levels 10.2 x 10.2 centimeters