Learn More
Med Vet International Foam Dressing ComfortFoam™ Border 5 X 8 Inch With Border Waterproof Backing Silicone Adhesive Elbow / Heel Sterile, 43580, Each
Catalog No. 503043872
Encompass_Preferred
The 5 x 8-inch elbow/heel DermaRite ComfortFoam border facilitates faster and proper healing of wounds A multilayer design optimizes wound protection, thermal insulation and absorbency The skin-friendly adhesive foam dressing sticks well and removes easily A low-profile border improves adhesion and flexibility for application in difficult areas
Catalog No. 50-304-3872 Supplier Med Vet International Supplier No. 946559EA
The 5 x 8-inch elbow/heel DermaRite ComfortFoam border facilitates faster and proper healing of wounds A multilayer design optimizes wound protection, thermal insulation and absorbency The skin-friendly adhesive foam dressing sticks well and removes easily A low-profile border improves adhesion and flexibility for application in difficult areas