Learn More
Med Vet International Mesalt Sodium Chloride Impregnated Dressing, ¾ x 39 Inch, Sterile, 285280, 10/Box
Catalog No. 503043659
Encompass_Preferred
For wounds with medium to high levels of exudate Stimulates wound cleansing Easy to use Sterile Ribbon is 0.75 inch wide and 39 inches long
Catalog No. 50-304-3659 Supplier Med Vet International Supplier No. 1087157BX
For wounds with medium to high levels of exudate Stimulates wound cleansing Easy to use Sterile Ribbon is 0.75 inch wide and 39 inches long