Learn More
Med Vet International McKesson Octagonal Sterile Dressing with Frame-Style Delivery, 6 x 8 Inch, Transparent, 4987, 80/Case
Catalog No. 503045203
Encompass_Preferred
Transparent Film Dressing McKesson Octagon 6 X 8 Inch Frame Style Delivery Without Label Sterile Sterile dressing is semi-permeable and water resistant Intended to provide a moist environment for granulating or superficial wounds Transparent film allows the site to be visually monitored Intended for single-use applications
Catalog No. 50-304-5203 Supplier Med Vet International Supplier No. 886410CS
Transparent Film Dressing McKesson Octagon 6 X 8 Inch Frame Style Delivery Without Label Sterile Sterile dressing is semi-permeable and water resistant Intended to provide a moist environment for granulating or superficial wounds Transparent film allows the site to be visually monitored Intended for single-use applications