Learn More
Med Vet International McKesson Octagonal Sterile Dressing with Frame-Style Delivery, 4 x 4-3/4 Inch, Transparent, 4986, Each
Catalog No. 503045248
Encompass_Preferred
Transparent Film Dressing McKesson Octagon 4 X 4-3/4 Inch Frame Style Delivery Without Label Sterile Sterile dressing is semi-permeable and water resistant Intended to provide a moist environment for granulating or superficial wounds Transparent film allows the site to be visually monitored Intended for single-use applications
Catalog No. 50-304-5248 Supplier Med Vet International Supplier No. 886409EA
Transparent Film Dressing McKesson Octagon 4 X 4-3/4 Inch Frame Style Delivery Without Label Sterile Sterile dressing is semi-permeable and water resistant Intended to provide a moist environment for granulating or superficial wounds Transparent film allows the site to be visually monitored Intended for single-use applications