Learn More
Med Vet International Vaseline™ Petrolatum Impregnated Dressing, 3 x 9 Inch, Sterile, 8884413605, 50/Box
Catalog No. 503043183
Encompass_Preferred
Nonadhesive Clings and conforms to all body contours Petrolatum impregnant remains moist-nontoxic, nonsensitizing and non-irritating Peelable foil packs
Catalog No. 50-304-3183 Supplier Med Vet International Supplier No. 163164BX
Nonadhesive Clings and conforms to all body contours Petrolatum impregnant remains moist-nontoxic, nonsensitizing and non-irritating Peelable foil packs