Learn More
Med Vet International DermaRite Sterile Gauze Adhesive Dressing, 4 x 10 Inch, 11410, Each
Catalog No. 503045359
Encompass_Preferred
This sterile wound dressing is soft, flexible and non-restrictive The first layer features a low adherent, lint-free material designed to protect the wound surface The second layer is super absorbent, removing released wound fluid The third layer is a nonwoven tape with a strong adhesive to keep the dressing secure and maintain a moist environment 4- x 10-inch DermaRite dressing is suitable for use on all skin types It comes individually wrapped to prevent contamination
Catalog No. 50-304-5359 Supplier Med Vet International Supplier No. 946446EA
This sterile wound dressing is soft, flexible and non-restrictive The first layer features a low adherent, lint-free material designed to protect the wound surface The second layer is super absorbent, removing released wound fluid The third layer is a nonwoven tape with a strong adhesive to keep the dressing secure and maintain a moist environment 4- x 10-inch DermaRite dressing is suitable for use on all skin types It comes individually wrapped to prevent contamination