Learn More
Med Vet International Vaseline Petrolatum Impregnated Dressing, 1 x 8 Inch, Sterile, 8884417601, 200/Case
Catalog No. 503044407
Encompass_Preferred
Fine mesh, absorbent gauze impregnated with white petrolatum Non-adherent Clings and conforms to all body contours Petrolatum impregnant remains moist-nontoxic, nonsensitizing and non-irritating Sterile 1 x 8 inches
Catalog No. 50-304-4407 Supplier Med Vet International Supplier No. 167694CS
Fine mesh, absorbent gauze impregnated with white petrolatum Non-adherent Clings and conforms to all body contours Petrolatum impregnant remains moist-nontoxic, nonsensitizing and non-irritating Sterile 1 x 8 inches