Learn More
Med Vet International Solosite Hydrogel Dressing, 3-ounce tube, 449600, 12/Case
Catalog No. 503043237
Encompass_Preferred
Solosite hydrogel wound dressing is composed of absorbent polymers which absorb the mass of wound drainage to keep the affected region clean; provides temporary relief from pain for up to 6 hours Gentle formula of the dressing gel assists in autolytic debridement; enzymes and moisture break down dead tissue; non-sensitizing; won't cause reactions in newly forming tissue Helps in cooling down the wounded area; commonly used on minor burns, lacerations, skin tears, diabetic foot ulcers and surgical incisions Wound gel with preservatives contains allantoin; softens the top layer of skin, helping the wound shed dead cells and reducing scar tissue 85-gram tube has a flip-top cap
Catalog No. 50-304-3237 Supplier Med Vet International Supplier No. 224395CS
Solosite hydrogel wound dressing is composed of absorbent polymers which absorb the mass of wound drainage to keep the affected region clean; provides temporary relief from pain for up to 6 hours Gentle formula of the dressing gel assists in autolytic debridement; enzymes and moisture break down dead tissue; non-sensitizing; won't cause reactions in newly forming tissue Helps in cooling down the wounded area; commonly used on minor burns, lacerations, skin tears, diabetic foot ulcers and surgical incisions Wound gel with preservatives contains allantoin; softens the top layer of skin, helping the wound shed dead cells and reducing scar tissue 85-gram tube has a flip-top cap