Learn More
Med Vet International McKesson Absorbent Gelling Fiber Dressing, 4 x 4-3/4 Inch, 87400, Each
Catalog No. 503044822
Encompass_Preferred
Absorbent Gelling Fiber Dressing McKesson Carboxymethyl Cellulose (CMC) 4 X 4-3/4 Inch Gelling fiber dressings may be used during the healing process of decubitus ulcers (pressure ulcers); venous and arterial leg ulcers; diabetic ulcers; graft and donor sites; post-operative surgical wounds; superficial and partial thickness burns and cavity wounds Highly absorbent Excellent wet and dry strength Supports moist wound environment for faster healing and low risk of maceration 4 x 4-3/4 inches
Catalog No. 50-304-4822 Supplier Med Vet International Supplier No. 1138627EA
Absorbent Gelling Fiber Dressing McKesson Carboxymethyl Cellulose (CMC) 4 X 4-3/4 Inch Gelling fiber dressings may be used during the healing process of decubitus ulcers (pressure ulcers); venous and arterial leg ulcers; diabetic ulcers; graft and donor sites; post-operative surgical wounds; superficial and partial thickness burns and cavity wounds Highly absorbent Excellent wet and dry strength Supports moist wound environment for faster healing and low risk of maceration 4 x 4-3/4 inches