Learn More
Med Vet International DermaRite Gauze Adhesive Dressing, White, Sterile, 4 X 8 inch, 11480, 25/Box
Catalog No. 503043048
Encompass_Preferred
This sterile wound dressing is soft, flexible and non-restrictive The first layer features a low adherent, lint-free material designed to protect the wound surface The second layer is super absorbent, removing released wound fluid The third layer is a nonwoven tape with a strong adhesive to keep the dressing secure and maintain a moist environment The 4 x 8 inch DermaRite adhesive gauze dressing is suitable for use on all skin types It comes individually wrapped to prevent contamination
Catalog No. 50-304-3048 Supplier Med Vet International Supplier No. 946445BX
This sterile wound dressing is soft, flexible and non-restrictive The first layer features a low adherent, lint-free material designed to protect the wound surface The second layer is super absorbent, removing released wound fluid The third layer is a nonwoven tape with a strong adhesive to keep the dressing secure and maintain a moist environment The 4 x 8 inch DermaRite adhesive gauze dressing is suitable for use on all skin types It comes individually wrapped to prevent contamination