Learn More
Med Vet International Aquacel Extra™ Gelling Fiber Wound Dressing, 2 × 2 Inch, Sterile, 420671, Each
Catalog No. 503043628
Encompass_Preferred
This hydrofiber wound dressing is nine times stronger and 39% more absorbent than a standard hydrofiber dressing Aquacel 2 × 2 inch wound dressing is made with two-dimensional strengthening fibers for high strength and durability It gels on contact with fluid for high absorbency It creates a moist environment that helps skin heal faster Use the Aquacel Extra hydrofiber dressing on moderate to highly exuding wounds
Catalog No. 50-304-3628 Supplier Med Vet International Supplier No. 785781EA
This hydrofiber wound dressing is nine times stronger and 39% more absorbent than a standard hydrofiber dressing Aquacel 2 × 2 inch wound dressing is made with two-dimensional strengthening fibers for high strength and durability It gels on contact with fluid for high absorbency It creates a moist environment that helps skin heal faster Use the Aquacel Extra hydrofiber dressing on moderate to highly exuding wounds